Proteintech
Manufacturer Code:510752AP150UL
Catalog # 50-173-5893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: interferon alpha induced transcriptional activator; ISGF 3 P113 STAT113 STAT2; p113; Signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa
Gene Aliases: IMD44; ISGF-3; P113; STAT113; STAT2
UniProt ID: (Human) B4DLC7
Entrez Gene ID: (Human) 6773
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.