Proteintech
Manufacturer Code:226731AP150UL
Catalog # 50-173-3919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | STARS/ABRA Fusion Protein Ag18456 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABRA; ABRA STARS; actin-binding Rho activating protein; Actin-binding Rho-activating protein; STARS; Striated muscle activator of Rho-dependent signaling
Gene Aliases: ABRA; C130068O12Rik; STARS
UniProt ID: (Human) Q8N0Z2, (Mouse) Q8BUZ1
Entrez Gene ID: (Human) 137735, (Mouse) 223513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.