Invitrogen
Manufacturer Code:PA559241
Catalog # PIPA559241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human STARD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: StAR-related lipid transfer (START) domain containing 5; StAR-related lipid transfer domain containing 5; StAR-related lipid transfer protein 5; StAR-related lipid transfer protein 5 START domain-containing protein 5 StARD5 STARD5; StARD5; START domain containing 5; START domain-containing protein 5
Gene Aliases: STARD5
UniProt ID: (Human) Q9NSY2
Entrez Gene ID: (Human) 80765
Molecular Function: membrane traffic protein transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.