OriGene
Manufacturer Code:CF810511
Catalog # 50-167-0125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8B4 |
Immunogen | Full length human recombit protein of human STARD4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: StAR related lipid transfer domain containing 4; StAR-related lipid transfer (START) domain containing 4; StAR-related lipid transfer domain containing 4; StAR-related lipid transfer protein 4; StARD4; START domain containing 4, sterol regulated; START domain-containing protein 4
Gene Aliases: STARD4
UniProt ID: (Human) Q96DR4
Entrez Gene ID: (Human) 134429
Molecular Function:
membrane traffic protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.