Invitrogen
Manufacturer Code:PA567611
Catalog # PIPA567611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human STARD4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: StAR-related lipid transfer (START) domain containing 4; StAR-related lipid transfer domain containing 4; StAR-related lipid transfer protein 4; StARD4; START domain containing 4, sterol regulated; START domain-containing protein 4
Gene Aliases: 4632419C16Rik; 9030213J02Rik; AA517649; AW324468; STARD4
UniProt ID: (Human) Q96DR4, (Mouse) Q99JV5
Entrez Gene ID: (Human) 134429, (Mouse) 170459
Molecular Function:
membrane traffic protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.