Novus Biologicals
Manufacturer Code:NBP184508
Catalog # NBP184508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:MECCDVPAETLYDVLHDIEYRKKWDSNVIETFDIARLTVNADVGYYSWRCPKPLKNRDVITLRSWLPMGADYIIMNYSVKHPKYPPRKDLVRAVSIQTGYL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-28; Antigen NY-CO-28 CGI-52 MGC14401 NY-CO-28 PCTP2 PCTP-L PCTP-like protein Serologically defined colon cancer antigen 28SDCCAG28 StARD10 StAR-related lipid transfer (START) domain containing 10 StAR-related lipid transfer protein 10 START domain containing 10 START domain-containing protein 10; PCTP-L; PCTP-like protein; Serologically defined colon cancer antigen 28; StAR-related lipid transfer (START) domain containing 10; StAR-related lipid transfer domain containing 10; StAR-related lipid transfer protein 10; StARD10; START domain containing 10; START domain-containing protein 10
Gene Aliases: CGI-52; NY-CO-28; PCTP2; SDCCAG28; STARD10
UniProt ID: (Human) Q9Y365
Entrez Gene ID: (Human) 10809
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.