Invitrogen
Manufacturer Code:PA521687
Catalog # PIPA521687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 15 and 268 of STAR (Uniprot IDNo.P49675) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cholesterol trafficker; Luteinizing hormone-induced protein; mitochondrial steroid acute regulatory protein; StAR; StAR related lipid transfer (START) domain containing 1; StAR-related lipid transfer (START) domain containing 1; StARD1; STARD1 START domain containing 1 START domain-containing protein 1 StAR-related lipid transfer (START) domain containing 1 cholesterol trafficker mitochondrial steroid acute regulatory protein steroid acute regulatory protein steroidogenic acute regulator steroidogenic acute regulatory protein mitochondrial; START domain containing 1; START domain-containing protein 1; steroid acute regulatory protein; steroidogenic acute regulator; Steroidogenic acute regulatory protein, mitochondrial; testis secretory sperm-binding protein Li 241mP
Gene Aliases: AV363654; D8Ertd419e; STAR; STARD1
UniProt ID: (Human) P49675, (Mouse) P51557
Entrez Gene ID: (Human) 6770, (Mouse) 20845
Molecular Function:
membrane traffic protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.