Invitrogen
Manufacturer Code:PA511545
Catalog # PIPA511545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 41-70 amino acids from the N-terminal region of human BRDG 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCR downstream signaling 1; BCR downstream-signaling protein 1; Docking protein BRDG1; Signal-transducing adaptor protein 1; STAP-1; STAP-1 Stem cell adaptor protein 1 BCR downstream-signaling protein 1 Docking protein BRDG1; Stem cell adaptor protein 1; stem cell adaptor protein STAP-1
Gene Aliases: AI586015; BRDG1; STAP-1; STAP1
UniProt ID: (Human) Q9ULZ2, (Mouse) Q9JM90
Entrez Gene ID: (Human) 26228, (Mouse) 56792
Molecular Function:
protein kinase receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.