R&D Systems
Manufacturer Code:FAB4626N100UG
Catalog # FAB4626N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 418714 |
Immunogen | NS0 mouse myeloma cell line transfected with human STAMP2 Met1-His459 Accession # EAW76907 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: EC 1.16.1 EC 1.16.1.- FLJ23153 Six-transmembrane epithelial antigen of prostate 4 SixTransMembrane protein of prostate 2 STAMP2DKFZp666D049 STEAP family member 4 STEAP4 TIARP TIARPsix transmembrane prostate protein 2 TNFAIP9 Tumor necrosis factor alpha-induced protein 9metalloreductase STEAP4 tumor necrosis-alpha-induced adipose-related protein; Metalloreductase STEAP4; six transmembrane prostate protein 2; Six-transmembrane epithelial antigen of prostate 4; SixTransMembrane protein of prostate 2; STEAP family member 4; Tumor necrosis factor, alpha-induced protein 9; tumor necrosis-alpha-induced adipose-related protein
Gene Aliases: STAMP2; STEAP4; TIARP; TNFAIP9
UniProt ID: (Human) Q687X5
Entrez Gene ID: (Human) 79689
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.