Invitrogen
Manufacturer Code:PA529187
Catalog # PIPA529187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 302 of Human STAMBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMSH STAM-binding protein associated molecule with the SH3 domain of STAM endosome-associated ubiquitin isopeptidase; Associated molecule with the SH3 domain of STAM; Endosome-associated ubiquitin isopeptidase; STAM-binding protein; testicular secretory protein Li 54
Gene Aliases: 5330424L14Rik; 5730422L11Rik; AMSH; AW107289; MICCAP; mKIAA4198; STAMBP
UniProt ID: (Human) O95630, (Mouse) Q9CQ26
Entrez Gene ID: (Human) 10617, (Mouse) 70527
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.