Invitrogen
Manufacturer Code:PA5116995
Catalog # PIPA5116995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 120-210 of human STAM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686J2352 HSE1 homolog RGD1564499 Signal transducing adapter molecule 1 signal transducing adaptor molecule signal transducing adaptor molecule (SH3 domain and ITAM motif) 1 Stam STAM1 STAM-1 STAT91; HSE1 homolog; Signal transducing adapter molecule 1; signal transducing adaptor molecule (SH3 domain and ITAM motif) 1; STAM-1
Gene Aliases: RGD1564499; STAM; STAM-1; STAM1
UniProt ID: (Human) Q92783, (Mouse) P70297
Entrez Gene ID: (Human) 8027, (Mouse) 20844, (Rat) 498798
Molecular Function:
membrane traffic protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.