Invitrogen
Manufacturer Code:PA585641
Catalog # PIPA585641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human SA2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA517O1.1 Antibody SA2 Antibody stromal antigen 2 Antibody SA-2 Antibody SCC3B Antibody STAG2 Antibody; Cohesin subunit SA-2; SCC3 homolog 2; Stromal antigen 2
Gene Aliases: bA517O1.1; SA-2; SA2; SCC3B; STAG2
UniProt ID: (Human) Q8N3U4
Entrez Gene ID: (Human) 10735
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.