Invitrogen
Manufacturer Code:PA519217
Catalog # PIPA519217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SRGSTVRSKKSKPST) corresponding to the internal amino acids of STAG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cohesin subunit SA-2; nuclear protein SA2; SCC3 homolog 2; STAG2 stromal antigen 2 DKFZp686P168 DKFZp781H1753 FLJ25871 SA-2 SA2 bA517O1.1 OTTHUMP00000024338 OTTHUMP00000043514 OTTHUMP00000043515 OTTHUMP00000081754 OTTHUMP00000081755 SCC3 homolog 2; Stromal antigen 2
Gene Aliases: 9230105L23Rik; B230112I07Rik; bA517O1.1; SA-2; SA2; SAP2; SCC3B; STAG2
UniProt ID: (Human) Q8N3U4, (Mouse) Q6NZN7
Entrez Gene ID: (Human) 10735, (Mouse) 20843
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.