Novus Biologicals
Manufacturer Code:NB10060946
Catalog # NB10060946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-REPKRLRPEDS corresponding to internal region according to NP_001036214.1 NP_001036215.1 NP_001036216.1 NP_006594.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bA517O1.1 cohesin subunit SA-2 DKFZp686P168 SA-2 SA2FLJ25871 SCC3 homolog 2 stromal antigen 2DKFZp781H1753; Cohesin subunit SA-2; SCC3 homolog 2; Stromal antigen 2
Gene Aliases: bA517O1.1; SA-2; SA2; SCC3B; STAG2
UniProt ID: (Human) Q8N3U4
Entrez Gene ID: (Human) 10735
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.