Novus Biologicals
Manufacturer Code:NB100301
Catalog # NB100301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 31 amino acid synthetic peptide which represented a portion of human Stromal Antigen 2 encoded in part by exon 31 and 32 (LocusLink ID 10735). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bA517O1.1 cohesin subunit SA-2 DKFZp686P168 SA-2 SA2FLJ25871 SCC3 homolog 2 stromal antigen 2DKFZp781H1753; Cohesin subunit SA-2; SCC3 homolog 2; Stromal antigen 2
Gene Aliases: bA517O1.1; SA-2; SA2; SCC3B; STAG2
UniProt ID: (Human) Q8N3U4
Entrez Gene ID: (Human) 10735
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.