Abnova
Manufacturer Code:PAB20556
Catalog # 89-122-061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human ST8SIA6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant ST8SIA6.
For Research Use Only
Protein Aliases: Alpha-2,8-sialyltransferase 8F; alpha-2,8-sialyltransferase 8F variant 1; alpha-2,8-sialyltransferase 8F variant 2; alpha-2,8-sialyltransferase 8F variant 3; SIA8F; Sialyltransferase 8F; sialyltransferase 8F (alpha-2, 8-sialyltransferase); Sialyltransferase St8Sia VI; sialytransferase St8Sia VI; SIAT8-F; SIAT8F; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 6; ST8SIA-VI; ST8SiaVI
Gene Aliases: SIA8F; SIAT8F; ST8SIA-VI; ST8SIA6
UniProt ID: (Human) P61647
Entrez Gene ID: (Human) 338596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.