Abnova
Manufacturer Code:H00007903M03
Catalog # 89-002-728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H5 |
Immunogen | ST8SIA4 (AAH27866 1 a.a. ~ 168 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ST8SIA4.
For Research Use Only
Protein Aliases: Alpha-2,8-sialyltransferase 8D; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; MGC34450; MGC61459; Polysialyltransferase-1; PST; PST1; sialyltransferase 8 (alpha-2, 8-polysialytransferase) D; Sialyltransferase 8D; Sialyltransferase St8Sia IV; sialytransferase St8Sia IV; SIAT8-D; SIAT8D; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 4; ST8SIA-IV; ST8SiaIV
Gene Aliases: PST; PST1; SIAT8D; ST8SIA-IV; ST8SIA4
UniProt ID: (Human) Q92187
Entrez Gene ID: (Human) 7903
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.