Invitrogen
Manufacturer Code:PA593199
Catalog # PIPA593199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 20-168 of human ST8SIA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-2,8-sialyltransferase 8D; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; polysialyltransferase; Polysialyltransferase-1; sialyltransferase 8 (alpha-2 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2, 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-sialytransferase) D; Sialyltransferase 8D; Sialyltransferase St8Sia IV; sialytransferase St8Sia IV; SIAT8-D; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 4; ST8SiaIV
Gene Aliases: PST; PST1; SIAT8D; ST8SIA-IV; ST8SIA4
UniProt ID: (Human) Q92187
Entrez Gene ID: (Human) 7903, (Rat) 116696
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.