Invitrogen
Manufacturer Code:PA569551
Catalog # PIPA569551
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of rat ST8SIA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-2,8-polysialyltransferase IV; Alpha-2,8-sialyltransferase 8D; alpha-2,8-sialyltransferase ST8Sia IV; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase-like protein; N-glycan alpha 2,8-sialyltransferase; polysialyltransferase; Polysialyltransferase-1; PST PST1 SIAT8D ST8SIA-IV CMP-N-acetylneuraminate-poly-alpha-2 8-sialyl transferase CMP-N-acetylneuraminate-poly-alpha-2 8-sialyltransferase SIAT8-D ST8SiaIV alpha-2 8-sialyltransferase 8D polysialyltransferase-1 sialyltransferase 8 (alpha-2 8-polysialytransferase) D sialyltransf; Sia8D; sialyltransferase 8 (alpha-2 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2, 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-sialyltransferase) D; sialyltransferase 8 (alpha-2, 8-sialytransferase) D; Sialyltransferase 8D; Sialyltransferase St8Sia IV; sialytransferase St8Sia IV; SIAT8-D; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 4; ST8Sia IV; ST8SiaIV
Gene Aliases: PST; PST-1; PST1; siat 8D; SIAT8-D; SIAT8D; ST8SIA-IV; ST8SIA4; ST8SiaIV
UniProt ID: (Human) Q92187, (Bovine) Q6ZXC9, (Mouse) Q64692
Entrez Gene ID: (Human) 7903, (Rabbit) 100350253, (Bovine) 407768, (Pig) 641356, (Horse) 100064780, (Dog) 488886, (Guinea pig) 100716689, (Mouse) 20452, (Rat) 116696, (Zebrafish) 553211
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.