Invitrogen
Manufacturer Code:PA525508
Catalog # PIPA525508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 312-340 amino acids from the C-terminal region of human SIA8C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-2,8-sialyltransferase 8C; Alpha-2,8-sialyltransferase III; Sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase; Sialyltransferase 8C; sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase); Sialyltransferase St8Sia III; sialytransferase St8Sia III; SIAT8-C; SIAT8C ST8SiaIII SIAT8-C alpha-28-sialyltransferase 8C alpha-28-sialyltransferase III sia-alpha-23-Gal-beta-14-GlcNAc-R:alpha 28-sialyltransferase sialyltransferase 8C (alpha23Galbeta14GlcNAcalpha 28-sialyltransferase) sialytransferase St8Sia III; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 3; ST8SiaIII
Gene Aliases: SIAT8C; ST8SIA3; ST8SiaIII
UniProt ID: (Human) O43173
Entrez Gene ID: (Human) 51046
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.