Proteintech
Manufacturer Code:180041AP150UL
Catalog # 50-173-1930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ST8SIA3 Fusion Protein Ag12497 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 28 sialyltransferase 8C Sialyltransferase 8C Sialytransferase St8Sia III SIAT8 C SIAT8C ST8SIA3 ST8SiaIII; Alpha-2,8-sialyltransferase 8C; Alpha-2,8-sialyltransferase III; GT3-synthase; Sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase; sialyltransferase 8 (alpha-2, 8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 2,8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 28-sialyltransferase) C; sialyltransferase 8 C; Sialyltransferase 8C; sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase); Sialyltransferase St8Sia III; sialytransferase St8Sia III; SIAT8-C; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 3; ST8SiaIII
Gene Aliases: AI847333; SIAT8C; ST8SIA3; ST8SiaIII
UniProt ID: (Human) O43173, (Mouse) Q64689
Entrez Gene ID: (Human) 51046, (Rat) 25547, (Mouse) 20451
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.