Invitrogen
Manufacturer Code:PA5113279
Catalog # PIPA5113279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Alpha-28-sialyltransferase 8B protein (24-160AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI323367 alpha-28-sialyltransferase Alpha-28-sialyltransferase 8B alpha-28-sialyltransferase 8B 1 BOS_20022 cb394 HsT19690 id:ibd5161 LOC100341648 MGC116854 MGC116857 N-glycan alpha 28-sialyltransferase Polysialic acid synthase sialyltransferase 8 sialyltransferase 8 (alpha-2 8-sialytransferase) B sialyltransferase 8 (alpha-2 8-sialyltransferase) B sialyltransferase 8 (alpha-2 8-sialytransferase) B Sialyltransferase 8B Sialyltransferase St8Sia II Sialyltransferase X sialytransferase St8Sia II SIAT 8B siat8 Siat8b SIAT8-B ST8 alpha-N-acetylneuraminate alpha-28-sialyltransferase 2 ST8 alpha-N-acetyl-neuraminide alpha-28-sialyltransferase 2 ST8 alpha-N-acetyl-neuraminide alpha-28-sialyltransferase 2-like ST8Sia II St8sia2 st8sia2 protein ST8SiaII ST8SIA-II STX; Alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; Polysialic acid synthase; sialyltransferase 8 (alpha-2, 8-sialyltransferase) B; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; Sialyltransferase 8B; Sialyltransferase St8Sia II; Sialyltransferase X; sialytransferase St8Sia II; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8SiaII; STX
Gene Aliases: AI323367; HsT19690; SIAT8B; ST8SIA-II; ST8SIA2; ST8SiaII; STX
UniProt ID: (Human) Q92186, (Mouse) O35696
Entrez Gene ID: (Human) 8128, (Mouse) 20450
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.