Proteintech
Manufacturer Code:197361AP150UL
Catalog # 50-173-2516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 28 sialyltransferase 8B HsT19690 Sialyltransferase 8B Sialyltransferase X Sialytransferase St8Sia II SIAT8 B SIAT8B ST8SIA II ST8SIA2 ST8SiaII STX; Alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; Polysialic acid synthase; sialyltransferase 8 (alpha-2 8-sialytransferase) B; sialyltransferase 8 (alpha-2, 8-sialyltransferase) B; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; Sialyltransferase 8B; Sialyltransferase St8Sia II; Sialyltransferase X; sialytransferase St8Sia II; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8Sia II; ST8SiaII; STX
Gene Aliases: AI323367; HsT19690; SIAT8B; ST8SIA-II; ST8SIA2; ST8SiaII; STX
UniProt ID: (Human) Q92186, (Rat) Q07977, (Mouse) O35696
Entrez Gene ID: (Human) 8128, (Rat) 117523, (Mouse) 20450
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.