EMD Millipore
Manufacturer Code:MABS1997100UG
Catalog # MABS1997100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HIC0-3C5 |
Immunogen | Trypsin-dissociated human pancreatic islet cells with low levels of exocrine and ductal cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Stable for 1 year at 2-8°C from date of receipt. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-N-acetylneuraminide alpha-28-sialyltransferaseEC: 2.4.99.8Alpha-28-sialyltransferase 8AGanglioside GD3 synthaseGanglioside GT3 synthaseSialyltransferase 8ASIAT8-ASialyltransferase St8Sia IST8SiaI
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.