Proteintech
Manufacturer Code:249181AP150UL
Catalog # 50-173-4616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ST8SIA1 Fusion Protein Ag20429 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 28 sialyltransferase 8A Ganglioside GD3 synthase Ganglioside GT3 synthase GD3S Sialyltransferase 8A Sialytransferase St8Sia I SIAT8 SIAT8 A SIAT8A ST8SIA1 ST8SiaI; alpha-2, 8-sialyltransferase; Alpha-2,8-sialyltransferase 8A; Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase; disialoganglioside (GD3) synthase; Ganglioside GD3 synthase; Ganglioside GT3 synthase; ganglioside-specific alpha-2,8-polysialyltransferase; GD3 synthase; sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3 synthase) A; sialyltransferase 8 A; Sialyltransferase 8A; sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, GD3 synthase); Sialyltransferase St8Sia I; sialytransferase St8Sia I; SIAT8-A; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 1; ST8Sia I; ST8SiaI
Gene Aliases: 9330109E03Rik; GD3S; Sia-T; SIAT8; SIAT8-A; SIAT8A; ST8SIA1; ST8SiaI
UniProt ID: (Human) Q92185, (Mouse) Q64687
Entrez Gene ID: (Human) 6489, (Mouse) 20449
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.