R&D Systems
Manufacturer Code:AF6590SP
Catalog # AF6590SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human alpha-28-Sialyltransferase 8B/ST8SIA2 Asp24-Thr375 Accession # Q92186 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; alpha-28-sialyltransferase 8B alpha-28-sialyltransferase 8B 1 EC 2.4.99 EC 2.4.99.- HsT19690 MGC116854 MGC116857 sialyltransferase 8 (alpha-2 8-sialytransferase) B Sialyltransferase 8B Sialyltransferase X Sialytransferase St8Sia II SIAT8B SIAT8-B ST8 alpha-N-acetyl-neuraminide alpha-28-sialyltransferase 2 ST8SiaII ST8SIA-II STX STXST8SiaII; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; Sialyltransferase 8B; Sialyltransferase St8Sia II; Sialyltransferase X; sialytransferase St8Sia II; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8SiaII; STX
Gene Aliases: HsT19690; SIAT8B; ST8SIA-II; ST8SIA2; STX
UniProt ID: (Human) Q92186
Entrez Gene ID: (Human) 8128
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.