Novus Biologicals
Manufacturer Code:NBP180750
Catalog # NBP180750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RLPNEKEIVQGVLQQGTAWRRNQTAARAFRKQMEDCCDPAHLFAMTKMNSPMGKSMWYDGEFLYSFTIDNSTYSLFPQATPFQLPLKKCAVVGNGGILKKSGCGRQIDEANFVMRC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-2,8-sialyltransferase 8A; Alpha-28-sialyltransferase 8A alpha-N-acetylneuraminide alpha-28-sialyltransferase disialoganglioside (GD3) synthase Ganglioside GD3 synthase Ganglioside GT3 synthase ganglioside-specific alpha-28-polysialyltransferase GD3 synthase) GD3S sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-28-sialytransferase GD3synthase) A Sialyltransferase 8A sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-28-sialyltransferase Sialytransferase St8Sia I SIAT8A SIAT8-A SIAT8EC 2.4.99.8 ST8 alpha-N-acetyl-neuraminide alpha-28-sialyltransferase 1 ST8SiaI; Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase; disialoganglioside (GD3) synthase; Ganglioside GD3 synthase; Ganglioside GT3 synthase; ganglioside-specific alpha-2,8-polysialyltransferase; sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3 synthase) A; Sialyltransferase 8A; sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, GD3 synthase); Sialyltransferase St8Sia I; sialytransferase St8Sia I; SIAT8-A; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 1; ST8SiaI
Gene Aliases: GD3S; SIAT8; SIAT8-A; SIAT8A; ST8SIA1; ST8SiaI
UniProt ID: (Human) Q92185
Entrez Gene ID: (Human) 6489
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.