Abnova
Manufacturer Code:H00030815K
Catalog # 89-202-327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ST6GALNAC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ST6GALNAC6 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)2,6-sialyltransferase member VI; GalNAc alpha-2,6-sialyltransferase VI; RP11-203J24.3 SIAT7F ST6GALNACVI; Sialyltransferase 7F; sialytransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialytransferase) F; SIAT7-F; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; ST6 -N-acetylgalactosaminide alpha-26-sialyltransferase 6; ST6 GalNAc alpha-2,6-sialyltransferase 6; ST6GalNAc VI; ST6GalNAcVI
Gene Aliases: SIAT7-F; SIAT7F; ST6GALNAC6; ST6GALNACVI; UNQ708/PRO1359
UniProt ID: (Human) Q969X2
Entrez Gene ID: (Human) 30815
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.