Invitrogen
Manufacturer Code:MA524336
Catalog # PIMA524336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 719508 |
Immunogen | CHO-derived recombit human SGalNAc alpha-26-sialyltransferase V/ST6GALNAC5 Gly30-Phe336 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase V; alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase E; GalNAc alpha-2,6-sialyltransferase V; GD1 alpha synthase; GD1 alpha synthase SIAT7E ST6GalNAcV alpha-N-acetylneuraminyl 23-betagalactosyl-13)-N-acetyl galactosaminidealpha-26-sialyltransferase E EC 2.4.99 EC 2.4.99.- EC 2.4.99.7 GalNAc alpha-26-sialyltransferase V GD1 alpha synthase MGC3184 sialyltransferase 7 (alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminide alpha-26-sialyltransferase) E sialyltransferase 7((alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminidealpha-26-sialyltransferase) E Sialyltransferase 7E SIAT7-E SIAT7Ealpha-N-acetylgalactosaminide alpha-26-sialyltransferase V ST6 (alpha-N-acetyl-neuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminidealpha-26-sialyltransferase 5 ST6 neuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminidealpha-26-sialyltransferase 5 ST6GalNAc V ST6GalNAcValpha-N-acetylgalactosaminide alpha-26-sialyltransferase 5; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) E; Sialyltransferase 7E; SIAT7-E; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6 GalNAc alpha-2,6-sialyltransferase 5; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6GalNAc V; ST6GalNAcV
Gene Aliases: SIAT7-E; SIAT7E; ST6GALNAC5; ST6GalNAcV
UniProt ID: (Human) Q9BVH7
Entrez Gene ID: (Human) 81849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.