Invitrogen
Manufacturer Code:PA5112276
Catalog # PIPA5112276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human ST6GALNAC5 fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI851940 Alpha-N-acetylgalactosaminide alpha-26-sialyltransferase 5 alpha-N-acetylgalactosaminide alpha-26-sialyltransferase V alpha-N-acetylneuraminyl 23-betagalactosyl-13)-N-acetyl galactosaminide alpha-26-sialyltransferase E BOS_3414 EC 2.4.99 EC 2.4.99.7 GalNAc alpha-26-sialyltransferase V GD1 alpha synthase LOC100344332 LOC100715749 MGC3184 sialyltransferase 7 ((alpha-N-acetylneuraminyl 23-betagalactosyl-13)-N-acetyl galactosaminide alpha-26-sialyltransferase) E sialyltransferase 7 ((alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminide alpha-26-sialyltransferase) E Sialyltransferase 7E Siat7e SIAT7-E ST6 (alpha-N-acetyl-neuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminide alpha-26-sialyltransferase 5 ST6 GalNAc alpha-26-sialyltransferase 5 ST6 N-acetylgalactosaminide alpha-26-sialyltransferase 5 ST6 neuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminide alpha-26-sialyltransferase 5 ST6GalNAc V St6galnac5 ST6GalNAcV; Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase V; alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase E; GalNAc alpha-2,6-sialyltransferase V; GD1 alpha synthase; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) E; Sialyltransferase 7E; SIAT7-E; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6 GalNAc alpha-2,6-sialyltransferase 5; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6GalNAc V; ST6GalNAcV
Gene Aliases: SIAT7-E; SIAT7E; ST6GALNAC5; ST6GalNAcV
UniProt ID: (Human) Q9BVH7
Entrez Gene ID: (Human) 81849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.