Invitrogen
Manufacturer Code:PA543295
Catalog # PIPA543295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ST6GALNAC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-2,6-sialyltransferase ST6GalNAc IV; Alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3-N-acetyl-galactosaminide alpha-2,6-sialyltransferase; beta-galactosamide alpha-2,6-sialyltransferase; GalNAc alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2, 6-sialyltransferase alpha2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase IV; Sia-alpha-2-3-Gal-beta-1-3-GalNAc GalNAc-alpha-2,6-sialyltransferase; Sialyltransferase 3C; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) D; Sialyltransferase 7D; sialyltransferase 7D ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase); SIAT3-C; SIAT7-D; ST6 (alpha-N-acetyl-neuraminyl-2 3-beta-galactosyl-1 3)-N-acetylgalactosaminide alpha-2 6-sialyltransferase 4 SIAT3C SIAT7D ST6GALNACIV; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 GalNAc alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 4; ST6GalNAc IV; ST6GalNAcIV
Gene Aliases: IV; SIAT3-C; SIAT3C; SIAT7-D; SIAT7B; SIAT7D; ST6GalNAc; st6GalNAc-IV; ST6GALNAC2; ST6GALNAC4; ST6GALNACIV
UniProt ID: (Human) Q9H4F1, (Mouse) Q9R2B6
Entrez Gene ID: (Horse) 100067121, (Bovine) 404124, (Rabbit) 100356933, (Dog) 609133, (Human) 27090, (Rat) 407764, (Guinea pig) 100725743, (Mouse) 20448
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.