Invitrogen
Manufacturer Code:PA531615
Catalog # PIPA531615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 83 and 295 of Human ST6GALNAC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3-N-acetyl-galactosaminide alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2, 6-sialyltransferase alpha2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase IV; Sialyltransferase 3C; Sialyltransferase 7D; sialyltransferase 7D ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase); SIAT3-C; SIAT3C SIAT7D ST6GALNACIV NeuAc-alpha-23-Gal-beta-13-GalNAc-alpha-2 6-sialyltransferase alpha26-sialyltransferase NeuAc-alpha-23-Gal-beta-13-GalNAc-alpha-26-sialyltransferase IV SIAT3-C SIAT7-D ST6GalNAc IV alpha-N-acetyl-neuraminyl-23-beta-galactosyl-13-N-acetyl-galactos; SIAT7-D; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 GalNAc alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 4; ST6GalNAc IV; ST6GalNAcIV
Gene Aliases: IV; SIAT3-C; SIAT3C; SIAT7-D; SIAT7D; ST6GalNAc; ST6GALNAC4; ST6GALNACIV
UniProt ID: (Human) Q9H4F1
Entrez Gene ID: (Human) 27090
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.