Invitrogen
Manufacturer Code:PA531353
Catalog # PIPA531353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 171 and 374 of Human ST6GALNAC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase; (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; GalNAc alpha-2,6-sialyltransferase II; SAITL1 SIAT7 SIAT7B SIATL1 ST6GalNAII STHM (alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminide B (alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminide alpha-26-sialyltransferase SIAT7-B ST6GalNAc II ST6GalNAcII alpha-N-acetylgalactosaminide alpha-26-; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; Sialyltransferase 7B; sialyltransferase-like 1; SIAT7-B; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 GalNAc alpha-2,6-sialyltransferase 2; ST6GalNAc II; ST6GALNAC2, alpha-2,6-sialyltransferase 2; ST6GalNAcII; SThM
Gene Aliases: SAITL1; SIAT7; SIAT7B; SIATL1; ST6GALNAC2; ST6GalNAII; STHM
UniProt ID: (Human) Q9UJ37
Entrez Gene ID: (Human) 10610
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.