Novus Biologicals
Manufacturer Code:NBP17921020UL
Catalog # NBP17921020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is St6galnac1. Peptide sequence GALLLLTALHLCDKVSAYGFITEGHQRFSDHYYDTSWKRLIFYINHDFRL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; alpha-N-acetylgalactosaminide alpha-26-sialyltransferase 1 EC 2.4.99.3 GalNAc alpha-2 6-sialyltransferase I long form GalNAc alpha-26-sialyltransferase I sialyltransferase 7((alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminidealpha-26-sialyltransferase) A Sialyltransferase 7A SIAT7A SIAT7-A ST6 (alpha-N-acetyl-neuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminidealpha-26-sialyltransferase 1 ST6GalNAc I ST6GalNAcIHSY11339 STYI; GalNAc alpha-2, 6-sialyltransferase I, long form; GalNAc alpha-2,6-sialyltransferase I; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) A; Sialyltransferase 7A; SIAT7-A; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6 GalNAc alpha-2,6-sialyltransferase 1; ST6GalNAc I; ST6GALNAC1, alpha-2,6-sialyltransferase 1; ST6GalNAcI
Gene Aliases: HSY11339; SIAT7A; ST6GALNAC1; ST6GalNAcI; STYI; UNQ543/PRO848
UniProt ID: (Human) Q9NSC7
Entrez Gene ID: (Human) 55808
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.