Invitrogen
Manufacturer Code:PA569666
Catalog # PIPA569666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse ST6GAL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2,6 sialyltransferase; Alpha 2,6-ST 2; alpha-2,6-sialyltransferase ST6Gal II; beta-galactosamide alpha-2,6-sialyltransferase II; Beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; sialyltransferase; Sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6Gal II; ST6Gal-II; ST6GalII
Gene Aliases: C230064G14Rik; KIAA1877; mKIAA1877; SIAT2; ST6Gal II; ST6Gal-II; ST6GAL2; ST6GalII
UniProt ID: (Bovine) A5D7T4, (Human) Q96JF0, (Mouse) Q76K27, (Rat) Q701R3
Entrez Gene ID: (Pig) 100512775, (Dog) 481315, (Rabbit) 100358153, (Bovine) 493992, (Horse) 100062033, (Human) 84620, (Mouse) 240119, (Rat) 301155, (Zebrafish) 403116
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.