Abnova
Manufacturer Code:H00084620K
Catalog # 89-203-686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ST6GAL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ST6GAL2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Alpha 2,6-ST 2; Beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; FLJ30711 FLJ37730 FLJ38334 KIAA1877 SIAT2 ST6GalII; sialyltransferase; Sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6Gal II; ST6GalII
Gene Aliases: KIAA1877; SIAT2; ST6GAL2; ST6GalII
UniProt ID: (Human) Q96JF0
Entrez Gene ID: (Human) 84620
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.