Invitrogen
Manufacturer Code:MA135289
Catalog # 01-671-251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | LN-1 |
Immunogen | Lymphocytes/SU-DHL4 cell line. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2,6-ST 1; B-cell antigen CD75; Beta-galactoside alpha-2,6-sialyltransferase 1; CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1; Sialyltransferase 1; sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase); Sialyltransferase 1 SIAT1 ST6GalI ST6N B-cell antigen CD75 CMP-N-acetylneuraminate beta-galactosamide alpha-2 6-sialyltransferase CMP-N-acetylneuraminate-beta-galactosamide-alpha-2 6-sialyltransferase 1 ST6Gal I alpha 2 6-ST 1 beta-galactoside alpha-2 6-sialyltransferase 1 sialyltransferase 1; ST6 beta-galactosamide alpha-2,6-sialyltranferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6Gal I; ST6GalI
Gene Aliases: SIAT1; ST6GAL1; ST6GalI; ST6N
UniProt ID: (Human) P15907
Entrez Gene ID: (Human) 6480
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.