Novus Biologicals
Manufacturer Code:NBP159959
Catalog # NBP159959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ST6GALNAC6(ST6 -N-acetylgalactosaminide alpha-26-sialyltransferase 6) The peptide sequence was selected from the C terminal of ST6GALNAC6. Peptide sequence YHYYEPKGPDECVTYIQNEHSRKGNHHRFITEKRVFSSWAQLYGIT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; alpha-N-acetylgalactosaminide alpha-26-sialyltransferase 6 CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)26-sialyltransferase member VI EC 2.4.99 EC 2.4.99.- EC 2.4.99.7 GalNAc alpha-26-sialyltransferase VI hST6GalNAc VI RP11-203J24.3 Sialyltransferase 7F sialytransferase 7 ((alpha-N-acetylneuraminyl 23-betagalactosyl-13)-N-acetylgalactosaminide alpha-26-sialytransferase) F SIAT7F SIAT7-F ST6 (alpha-N-acetyl-neuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminidealpha-26-sialyltransferase 6 ST6GalNAc VI ST6GalNAcVI; CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)2,6-sialyltransferase member VI; GalNAc alpha-2,6-sialyltransferase VI; Sialyltransferase 7F; sialytransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialytransferase) F; SIAT7-F; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; ST6 -N-acetylgalactosaminide alpha-26-sialyltransferase 6; ST6 GalNAc alpha-2,6-sialyltransferase 6; ST6GalNAc VI; ST6GalNAcVI
Gene Aliases: SIAT7-F; SIAT7F; ST6GALNAC6; ST6GALNACVI; UNQ708/PRO1359
UniProt ID: (Human) Q969X2
Entrez Gene ID: (Human) 30815
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.