Novus Biologicals
Manufacturer Code:NBP290715AF532
Catalog # NB076843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 25 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse ST6 Sialyltransferase 2/ST6GALNAC2 (P70277 Ser28-Arg373). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase; (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; (alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminide B alpha-N-acetylgalactosaminide alpha-26-sialyltransferase 2 EC 2.4.99.- FLJ45660 GalNAc alpha-26-sialyltransferase II SAITL1 sialyltransferase 7((alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminide B sialyltransferase 7((alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminidealpha-26-sialyltransferase) sialyltransferase 7((alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminidealpha-26-sialyltransferase) B Sialyltransferase 7B sialyltransferase-like 1 SIAT7 SIAT7B SIAT7-B SIATL1 ST6 (alpha-N-acetyl-neuraminyl-23-beta-galactosyl-13)-N-acetylgalactosaminidealpha-26-sialyltransferase 2 ST6GalNAc II ST6GalNAcII ST6GalNAII SThM STHM(alpha-N-acetylneuraminyl-23-beta-galactosyl-13)-N-acetyl galactosaminidealpha-26-sialyltransferase; Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; GalNAc alpha-2,6-sialyltransferase II; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; Sialyltransferase 7B; sialyltransferase-like 1; SIAT7-B; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 GalNAc alpha-2,6-sialyltransferase 2; ST6GalNAc II; ST6GALNAC2, alpha-2,6-sialyltransferase 2; ST6GalNAcII; SThM
Gene Aliases: SAITL1; SIAT7; SIAT7B; SIATL1; ST6GALNAC2; ST6GalNAII; STHM
UniProt ID: (Human) Q9UJ37
Entrez Gene ID: (Human) 10610
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.