R&D Systems
Manufacturer Code:AF7747
Catalog # AF7747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human ST6 Gal Sialyltransferase 2/ST6GAL2 Glu243-Ser529 Accession # Q96JF0 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2,6-ST 2; Alpha 26-ST 2 beta-galactoside alpha-26-sialyltransferase 2 beta-galactoside alpha-26-sialyltransferase II CMP-N-acetylneuraminate-beta-galactosamide-alpha-26-sialyltransferase 2 EC 2.4.99 EC 2.4.99.1 FLJ30711 FLJ37730 FLJ38334 hST6Gal II KIAA1877 KIAA1877St6gal2 Sialyltransferase 2 sialyltransferase 2 (monosialoganglioside sialyltransferase) SIAT2 ST6 beta-galactosamide alpha-26-sialyltranferase 2 ST6Gal II ST6GALII; Beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; sialyltransferase; Sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6Gal II; ST6GalII
Gene Aliases: KIAA1877; SIAT2; ST6GAL2; ST6GalII
UniProt ID: (Human) Q96JF0
Entrez Gene ID: (Human) 84620
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.