Novus Biologicals
Manufacturer Code:NBP2478900.1MG
Catalog # NBP247890A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | LN-1 |
Immunogen | Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2,6-ST 1; B-cell antigen CD75; B-cell antigen CD75 beta-galactoside alpha-26-sialyltransferase 1 CMP-N-acetylneuraminate beta-galactosamide alpha-26-sialyltransferase CMP-N-acetylneuraminate-beta-galactosamide-alpha-26-sialyltransferase 1 EC 2.4.99.1 MGC48859 Sialyltransferase 1 sialyltransferase 1 (beta-galactoside alpha-26-sialyltransferase) sialyltransferase 1 (beta-galactoside alpha-26-sialytransferase) SIAT1Alpha 26-ST 1 ST6 beta-galactosamide alpha-26-sialyltranferase 1 ST6Gal I ST6GalI ST6N; Beta-galactoside alpha-2,6-sialyltransferase 1; CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1; Sialyltransferase 1; sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase); ST6 beta-galactosamide alpha-2,6-sialyltranferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6Gal I; ST6GalI
Gene Aliases: SIAT1; ST6GAL1; ST6GalI; ST6N
UniProt ID: (Human) P15907
Entrez Gene ID: (Human) 6480
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.