R&D Systems
Manufacturer Code:MAB5924
Catalog # MAB5924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 396012 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ST6 Gal Sialytransferase 1/ST6GAL1 Lys27-Cys406 Accession # P15907 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2,6-ST 1; Alpha 26-ST B-cell antigen CD75 beta-galactoside alpha-26-sialyltransferase 1 CD75 CMP-N-acetylneuraminate beta-galactosamide alpha-26-sialyltransferase CMP-N-acetylneuraminate-beta-galactosamide-alpha-26-sialyltransferase 1 EC 2.4.99.1 MGC48859 Sialyltransferase 1 sialyltransferase 1 (beta-galactoside alpha-26-sialyltransferase) sialyltransferase 1 (beta-galactoside alpha-26-sialytransferase) SIAT1 SIAT1Alpha 26-ST 1 ST6 beta-galactosamide alpha-26-sialyltranferase 1 ST6Gal I ST6GalI ST6N; B-cell antigen CD75; Beta-galactoside alpha-2,6-sialyltransferase 1; CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1; Sialyltransferase 1; sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase); ST6 beta-galactosamide alpha-2,6-sialyltranferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6Gal I; ST6GalI
Gene Aliases: SIAT1; ST6GAL1; ST6GalI; ST6N
UniProt ID: (Human) P15907
Entrez Gene ID: (Human) 6480
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.