Invitrogen
Manufacturer Code:PA595166
Catalog # PIPA595166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human ST5 different from the related mouse and rat sequences by one amino acid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DENN domain-containing protein 2B; DENN/MADD domain containing 2B; HeLa tumor suppression 1; Suppression of tumorigenicity 5 protein; Suppression of tumorigenicity 5 protein DENN domain-containing protein 2B HeLa tumor suppression 1 ST5 DENND2B HTS1
Gene Aliases: 2010004M01Rik; 2610305K15Rik; Denn2b; DENND2B; HTS1; p126; ST5
UniProt ID: (Human) P78524, (Mouse) Q924W7
Entrez Gene ID: (Human) 6764, (Mouse) 76954, (Rat) 308944
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.