Invitrogen
Manufacturer Code:PA520748
Catalog # PIPA520748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide from near the carboxy terminus of human ST3gal6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-2,3-sialyltransferase VI; alpha2,3-sialyltransferase ST3Gal VI; CMP-NeuAc:beta-galactoside alpha-2,3-sialyltransferase VI; Sialyltransferase 10; sialyltransferase 10 (alpha-2,3-sialyltransferase VI); SIAT10 ST3GALVI CMP-NeuAc:beta-galactoside alpha-23-sialyltransferase VI ST3Gal VI alpha23-sialyltransferase sialyltransferase 10 (alpha-23-sialyltransferase VI) type 2 lactosamine alpha-23-sialyltransferase; ST3Gal VI; ST3GalVI; Type 2 lactosamine alpha-2,3-sialyltransferase
Gene Aliases: 1700023B24Rik; AI930218; AW552396; SIAT10; ST3GAL6; ST3GALVI
UniProt ID: (Human) Q9Y274, (Mouse) Q8VIB3
Entrez Gene ID: (Human) 10402, (Rat) 304023, (Mouse) 54613
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.