Novus Biologicals
Manufacturer Code:NBP2858300.1ML
Catalog # NB033759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human ST3GAL6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha2,3-sialyltransferase ST3Gal VI; alpha23-sialyltransferase CMP-NeuAc:beta-galactoside alpha-23-sialyltransferase VI EC 2.4.99 EC 2.4.99.- EC 2.4.99.9 Sialyltransferase 10 SIAT10 ST3 beta-galactoside alpha-23-sialyltransferase 6 ST3Gal VI ST3GalVI ST3GALVIsialyltransferase 10 (alpha-23-sialyltransferase VI) type 2 lactosamine alpha-23-sialyltransferase; CMP-NeuAc:beta-galactoside alpha-2,3-sialyltransferase VI; Sialyltransferase 10; sialyltransferase 10 (alpha-2,3-sialyltransferase VI); ST3Gal VI; ST3GalVI; Type 2 lactosamine alpha-2,3-sialyltransferase
Gene Aliases: SIAT10; ST3GAL6; ST3GALVI
UniProt ID: (Human) Q9Y274
Entrez Gene ID: (Human) 10402
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.