Invitrogen
Manufacturer Code:PA530289
Catalog # PIPA530289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 174 and 418 of Human ST3GAL5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2,3-sialyltransferase V; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; GM3-specific sialytransferase; Lactosylceramide alpha-2,3-sialyltransferase; mST3Gal V; SATI SIAT9 SIATGM3S ST3GalV CMP-NeuAc:lactosylceramide alpha-23-sialyltransferase GM3 synthase ST3Gal V alpha 23-sialyltransferase V ganglioside GM3 synthase lactosylceramide alpha-23-sialyltransferase sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-23-sialyltransferase G; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase); sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GalV
Gene Aliases: 3S-T; [a]2; GM3S; SATI; SIAT9; SIATGM3S; ST3GAL5; ST3GalV; UNQ2510/PRO5998
UniProt ID: (Human) Q9UNP4, (Mouse) O88829
Entrez Gene ID: (Human) 8869, (Mouse) 20454
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.