Abnova
Manufacturer Code:H00008869K
Catalog # 89-203-085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ST3GAL5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ST3GAL5 peptide using ARM Technology.
For Research Use Only
Protein Aliases: alpha 2,3-sialyltransferase V; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; Lactosylceramide alpha-2,3-sialyltransferase; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); SIAT9 SIATGM3S ST3GalV; ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GalV
Gene Aliases: SATI; SIAT9; SIATGM3S; ST3GAL5; ST3GalV; UNQ2510/PRO5998
UniProt ID: (Human) Q9UNP4
Entrez Gene ID: (Human) 8869
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.