Invitrogen
Manufacturer Code:PA5107282
Catalog # PIPA5107282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: IKQKPTTGLL AITLALHLCD LVHIAGFGYP DAYNKKQTIH YYEQITLKSM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2,3-sialyltransferase IV; Alpha 2,3-ST 4; alpha-3-N-acetylneuraminyltransferase; beta-galactoside alpha-2,3-sialyltransferase 4; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase; Gal-beta-1,4-GlcNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; N-acetyllactosaminide alpha-2,3-sialyltransferase; SAT-3; Sialyltransferase 4C; sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase); SIAT4-C; ST-4; ST3Gal IV; ST3GalA.2; ST3GalIV; STZ
Gene Aliases: CGS23; NANTA3; SAT3; SIAT4; SIAT4C; ST3GAL4; ST3GalIV; STZ
UniProt ID: (Human) Q11206
Entrez Gene ID: (Human) 6484
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.