Novus Biologicals
Manufacturer Code:NBP29378320UL
Catalog # NB029429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 27-267 of human ST3GAL4 (NP_006269.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2,3-sialyltransferase IV; Alpha 2,3-ST 4; Alpha 23-sialyltransferase IV Alpha 23-ST 4 Beta-galactoside alpha-23-sialyltransferase 4 CGS23FLJ46764 EC 2.4.99 EC 2.4.99.- EC 2.4.99.9 FLJ11867 gal-beta-14-GalNAc-alpha-23-sialyltransferase Gal-NAc6S NANTA3alpha-3-N-acetylneuraminyltransferase SAT3 SAT-3 Sialyltransferase 4C sialyltransferase 4C (beta-galactosidase alpha-23-sialytransferase) sialyltransferase 4C (beta-galactoside alpha-23-sialytransferase) SIAT4-C SIAT4CCMP-N-acetylneuraminate-beta-galactosamide-alpha-23-sialyltransferase 4 ST3 beta-galactoside alpha-23-sialyltransferase 4 ST3Gal IV ST3GalA.2 ST3GalIV ST-4 STZSIAT4; alpha-3-N-acetylneuraminyltransferase; beta-galactoside alpha-2,3-sialyltransferase 4; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase; Gal-beta-1,4-GlcNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; N-acetyllactosaminide alpha-2,3-sialyltransferase; SAT-3; Sialyltransferase 4C; sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase); SIAT4-C; ST-4; ST3Gal IV; ST3GalA.2; ST3GalIV; STZ
Gene Aliases: CGS23; NANTA3; SAT3; SIAT4; SIAT4C; ST3GAL4; ST3GalIV; STZ
UniProt ID: (Human) Q11206
Entrez Gene ID: (Human) 6484
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.