Invitrogen
Manufacturer Code:PA542912
Catalog # PIPA542912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ST3GAL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2,3-sialyltransferase III; Alpha 2,3-ST 3; alpha-2,3-sialyltransferase II; Beta-galactoside alpha-2,3-sialyltransferase 3; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase soluble form; Gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase; Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase; mental retardation, non-syndromic, autosomal recessive, 12; N-acetyllacosaminide alpha 2,3-sialyltransferase; N-acetyllactosaminide alpha-2,3-sialyltransferase; sialyltransferase (N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase 3; Sialyltransferase 6; sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase); sialyltransferase 6(N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase ST3Gal-III; ST3 beta-galactoside alpha-2 3-sialyltransferase 3 SIAT6 ST3GALII ST3GalIII ST3N MRT12; ST3Gal III; ST3GalIII; ST3N
Gene Aliases: EIEE15; MRT12; Siat3; SIAT6; ST3GAL3; ST3GALII; ST3GalIII; ST3N
UniProt ID: (Human) Q11203, (Mouse) P97325, (Rat) Q02734
Entrez Gene ID: (Rabbit) 100353106, (Bovine) 444859, (Human) 6487, (Dog) 610736, (Horse) 100053104, (Mouse) 20441, (Rat) 64445
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.